Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2-butyl-1-benzofuran-3-yl)-[3,5-diiodo-4-(3-pyrrolidin-1-ylpropoxy)phenyl]methanone | 782387-91-1 | 99.0% | C26H29I2NO3 | 10MG
SDP

Catalog No. 5000258382
Encompass_Preferred

ADTL-SA1215 is a small-molecule activator of SIRT3 used for research; it modulates autophagy in triple-negative breast cancer models and is supplied as a characterized solid reagent.

  • Specific activator of SIRT3 that modulates autophagy in triple-negative breast cancer.
  • High purity reported (98.96%).
  • Molecular formula C26H29I2NO3 and molecular weight 657.32.
  • Supplied as a 10 mg solid for research use.
  • Appearance: light brown to brown solid.
  • Identified by CAS 782387-91-1 and available structural identifiers.

Catalog No. 50-002-58382 Supplier Medchemexpress LLC Supplier No. HY13974210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ADTL-SA1215 is a small-molecule activator of SIRT3 used for research; it modulates autophagy in triple-negative breast cancer models and is supplied as a characterized solid reagent.

  • Specific activator of SIRT3 that modulates autophagy in triple-negative breast cancer.
  • High purity reported (98.96%).
  • Molecular formula C26H29I2NO3 and molecular weight 657.32.
  • Supplied as a 10 mg solid for research use.
  • Appearance: light brown to brown solid.
  • Identified by CAS 782387-91-1 and available structural identifiers.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.