Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butyl-3-[[2'-cyano-[1,1'-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4,4]non-1-en-4-one 97.0+%, TCI America™
SDP

Catalog No. B41431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41431G 1 g
B41435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41431G Supplier TCI America Supplier No. B41431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138401-24-8
Molecular Formula C25H27N3O
Molecular Weight (g/mol) 385.511
MDL Number MFCD06658242
InChI Key KWEQEHOPDHARIA-UHFFFAOYSA-N
PubChem CID 9843116
IUPAC Name 2-[4-[(2-butyl-4-oxo-1,3-diazaspiro[4.4]non-1-en-3-yl)methyl]phenyl]benzonitrile
SMILES CCCCC1=NC2(CCCC2)C(=O)N1CC3=CC=C(C=C3)C4=CC=CC=C4C#N

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 1 g
UN Number 3077
Synonym 4′C-[(2-Butyl-4-oxo-1,3-diazaspiro[4,4]non-1-en-3-yl)methyl]-[1,1′C-biphenyl]-2-carbonitrile
Formula Weight 385.51
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butyl-3-[[2′-cyano-[1,1′-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4,4]non-1-en-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.