Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran 98.0+%, TCI America™
SDP

Catalog No. B49385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49385G 5 g
B49381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49385G Supplier TCI America Supplier No. B49385G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1951-26-4
Molecular Formula C19H16I2O3
Molecular Weight (g/mol) 546.143
MDL Number MFCD02675787
InChI Key PNFMEGSMKIHDFZ-UHFFFAOYSA-N
Synonym (2-Butyl-3-benzofuranyl)(4-hydroxy-3,5-diiodophenyl)methanone
PubChem CID 74769
ChEBI CHEBI:79661
IUPAC Name (2-butyl-1-benzofuran-3-yl)-(4-hydroxy-3,5-diiodophenyl)methanone
SMILES CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)O)I

Specifications

Melting Point 145°C
Quantity 5 g
Formula Weight 546.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.