Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2-Butyl-5-nitrobenzofuran-3-yl)(4-hydroxyphenyl)methanone | 141645-16-1 | 99.6% | C19H17NO5 | 5 G
SDP

Catalog No. 5000434328
Encompass_Preferred

(2-Butyl-5-nitrobenzofuran-3-yl)(4-hydroxyphenyl)methanone is a drug intermediate for the synthesis of various active compounds. It is for research use only.

  • Drug intermediate for synthesis of various active compounds.
  • For research use only.
  • Molecular weight: 339.35
  • Formula: C19H17NO5
  • CAS number: 141645-16-1
  • Appearance: Solid
  • Color: Off-white to yellow
  • Purity: 99.6%
  • Store powder at -20°C for 3 years; 4°C for 2 years.
  • Store in solvent at -80°C for 6 months; -20°C for 1 month.

Catalog No. 50-004-34328 Supplier Medchemexpress LLC Supplier No. HY753165G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2-Butyl-5-nitrobenzofuran-3-yl)(4-hydroxyphenyl)methanone is a drug intermediate for the synthesis of various active compounds. It is for research use only.

  • Drug intermediate for synthesis of various active compounds.
  • For research use only.
  • Molecular weight: 339.35
  • Formula: C19H17NO5
  • CAS number: 141645-16-1
  • Appearance: Solid
  • Color: Off-white to yellow
  • Purity: 99.6%
  • Store powder at -20°C for 3 years; 4°C for 2 years.
  • Store in solvent at -80°C for 6 months; -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.