Learn More
Medchemexpress LLC (2-Butyl-5-nitrobenzofuran-3-yl)(4-hydroxyphenyl)methanone | 141645-16-1 | 99.6% | C19H17NO5 | 500 G

Supplier: Medchemexpress LLC HY75316500G
(2-Butyl-5-nitrobenzofuran-3-yl)(4-hydroxyphenyl)methanone is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. It is intended for research use only.
- Purity of 99.6%
- Appears as an off-white to yellow solid
- Molecular weight of 339.35
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.