Learn More
Medchemexpress LLC (2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone | 141627-42-1 | 99.6% | C20H19NO5 | 1 G
SDP
Catalog No. 5000433705
Encompass_Preferred
(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone is a drug intermediate used for the synthesis of various active compounds. It is for research use only and not intended for sale to patients.
  • Drug intermediate for synthesis of various active compounds
  • Purity of 99.6%
  • Appears as a yellow to brown solid
  • Stable under recommended storage conditions
Catalog No. 50-004-33705 Supplier Medchemexpress LLC Supplier No. HY753151G
(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone is a drug intermediate used for the synthesis of various active compounds. It is for research use only and not intended for sale to patients.
  • Drug intermediate for synthesis of various active compounds
  • Purity of 99.6%
  • Appears as a yellow to brown solid
  • Stable under recommended storage conditions