Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone | 141627-42-1 | 99.6% | C20H19NO5 | 25 G
SDP

Catalog No. 5000359023
Encompass_Preferred

(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone is a drug intermediate for the synthesis of various active compounds. It is for research use only.

  • Used as a drug intermediate for active compound synthesis
  • Molecular weight: 353.37
  • Chemical formula: C20H19NO5
  • Appearance: Solid, yellow to brown
  • Shipping: Room temperature in continental US
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-59023 Supplier Medchemexpress LLC Supplier No. 25GHY75315
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone is a drug intermediate for the synthesis of various active compounds. It is for research use only.

  • Used as a drug intermediate for active compound synthesis
  • Molecular weight: 353.37
  • Chemical formula: C20H19NO5
  • Appearance: Solid, yellow to brown
  • Shipping: Room temperature in continental US
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.