Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone | 141627-42-1 | 99.6% | C20H19NO5 | 5 G
SDP

Catalog No. 5000433951
Encompass_Preferred

(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone is a drug intermediate used for the synthesis of various active compounds. It is for research use only and not sold to patients.

  • Purity of 99.59%
  • Physical form: Solid
  • Color: Yellow to brown
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-33951 Supplier Medchemexpress LLC Supplier No. HY753155G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone is a drug intermediate used for the synthesis of various active compounds. It is for research use only and not sold to patients.

  • Purity of 99.59%
  • Physical form: Solid
  • Color: Yellow to brown
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.