Learn More
Medchemexpress LLC (2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone | 141627-42-1 | 99.6% | C20H19NO5 | 50 G
SDP
Catalog No. 5000434381
Encompass_Preferred
(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone is a chemical compound functioning as a drug intermediate for the synthesis of various active compounds. It is designated for research use only.
  • Functions as a drug intermediate for active compound synthesis
  • Intended for research use only
  • High purity of 99.6%
  • Appears as a solid
  • Color ranges from yellow to brown
Catalog No. 50-004-34381 Supplier Medchemexpress LLC Supplier No. HY7531550G
(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone is a chemical compound functioning as a drug intermediate for the synthesis of various active compounds. It is designated for research use only.
  • Functions as a drug intermediate for active compound synthesis
  • Intended for research use only
  • High purity of 99.6%
  • Appears as a solid
  • Color ranges from yellow to brown