Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butyl-6-phenylpyridine 97.0+%, TCI America™
SDP

Catalog No. B33141G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33141G Supplier TCI America Supplier No. B33141G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60272-71-1
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.308
InChI Key DSCUZYVYDVDBTP-UHFFFAOYSA-N
PubChem CID 4614438
IUPAC Name 2-butyl-6-phenylpyridine
SMILES CCCCC1=CC=CC(=N1)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 190°C
Quantity 1 g
Formula Weight 211.31
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Butyl-6-phenylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.