Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Butyl)triphenylphosphonium bromide, 96%

Catalog No. AAL0153206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0153206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0153206 Supplier Thermo Scientific Chemicals Supplier No. L0153206
  • $136.50
    $116.65 / Each of 1

    Save $19.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3968-92-1 | C22H24BrP | 399.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3968-92-1
Molecular Formula C22H24BrP
Molecular Weight (g/mol) 399.31
MDL Number MFCD00050236
InChI Key HKWAYXKPYJOYEL-UHFFFAOYNA-M
Synonym 2-butyl triphenylphosphonium bromide, sec-butyltriphenylphosphonium bromide, triphenyl sec-butyl phosphanium bromide, acmc-1cswd, butan-2-yl triphenyl phosphanium bromide, phosphonium, 1-methylpropyl triphenyl-, bromide 1:1
PubChem CID 3083826
IUPAC Name butan-2-yl(triphenyl)phosphanium;bromide
SMILES [Br-].CCC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 227°C to 237°C
Quantity 5 g
Beilstein 3580097
Sensitivity Hygroscopic
Formula Weight 399.32
Percent Purity 96%
Chemical Name or Material (2-Butyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-587-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.