Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butylbenzofuran 99.0+%, TCI America™
SDP

Catalog No. B182425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B182425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B182425G Supplier TCI America Supplier No. B182425G
Add to Cart
Add to Cart

Specifications

CAS 4265-27-4
Boiling Point 129°C
Molecular Formula C12H14O
Quantity 25 g
MDL Number MFCD00005851
Synonym 2-butylbenzofuran, benzofuran, 2-butyl, 2-n-butylbenzo b furan, 2-butyl-benzofuran, 2-n-butylbenzofuran, 2-butylbenzo b furan, butylbenzofuran, 2-n-butylbezofuran, butyl-2-benzofuran, 2-n-butylbenzofurane
InChI Key OVJKFJDEVKABNF-UHFFFAOYSA-N
SMILES CCCCC1=CC2=CC=CC=C2O1
IUPAC Name 2-butyl-1-benzofuran
Molecular Weight (g/mol) 174.24
PubChem CID 77951
Formula Weight 174.24
Percent Purity ≥99.0% (GC)
Chemical Name or Material 2-Butylbenzofuran
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.