Learn More
Echelon Biosciences Research Labs 2-C-Methyl-D-erythritol 2,4-cyclophosphate (cMEPP) 10 mg
SDP
Catalog No. 50143987
Encompass
2-C-Methyl-D-erythritol 2,4-cyclophosphate (cMEPP) is an intermediate in the non-mevalonate pathway. It is formed by from MECDP Synthase from CDP-MEP and converted to 1-hydroxy-2-methyl-2-buten-4-yl diphosphate (HDMAPP) by IspG (GcpE). This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-143-987 Supplier Echelon Biosciences Research Labs Supplier No. IM05410MG
Add to Cart
Add to Cart
Request a Quote
2-C-Methyl-D-erythritol 2,4-cyclophosphate (cMEPP) is an intermediate in the non-mevalonate pathway. It is formed by from MECDP Synthase from CDP-MEP and converted to 1-hydroxy-2-methyl-2-buten-4-yl diphosphate (HDMAPP) by IspG (GcpE). This item is non returnable