Learn More
Cayman Chemical 2'-C-Methyladenosine, Purity: ≥98%, CAS: 15397-12-3, 50 mg
SDP
Catalog No. 502964350
Encompass_Preferred
An adenosine analog; inhibits HCV replication in vitro (IC50 = 0.3 µM); converted intracellularly to adenosine triphosphate; inhibits growth of L. guyanensis in vitro (EC50 = 3 µM); eradicates L. guyanensis in vitro (10 µM)
Catalog No. 50-296-4350 Supplier Cayman Chemical Supplier No. 2306250 MG
An adenosine analog; inhibits HCV replication in vitro (IC50 = 0.3 µM); converted intracellularly to adenosine triphosphate; inhibits growth of L. guyanensis in vitro (EC50 = 3 µM); eradicates L. guyanensis in vitro (10 µM)