Learn More
Selleck Chemical LLC 2-C-Methylcytidine E6045-1G
SDP
Catalog No. 5000041074
Encompass_Preferred
2 -C-Methylcytidine (NM107) is a potent and selective inhibitor of Chronic hepatitis C virus (HCV) NS5B polymerase with an EC50 of 1 85 M in wild-type replicon cells It also effectively inhibits foot-and-mouth disease virus(FMDV) replication with an EC50 of 6 4 M Therefore it is a potent antiviral that targets both viral and host factors making it a promising option for HCV therapy and a potential prophylactic or therapeutic agent against FMDV outbreaks
Catalog No. 50-000-41074 Supplier Selleck Chemical LLC Supplier No. E60451G
2 -C-Methylcytidine (NM107) is a potent and selective inhibitor of Chronic hepatitis C virus (HCV) NS5B polymerase with an EC50 of 1 85 M in wild-type replicon cells It also effectively inhibits foot-and-mouth disease virus(FMDV) replication with an EC50 of 6 4 M Therefore it is a potent antiviral that targets both viral and host factors making it a promising option for HCV therapy and a potential prophylactic or therapeutic agent against FMDV outbreaks