Learn More
Medchemexpress LLC 2'-C-methyluridine | 31448-54-1 | MFCD02682960 | ≥97.0% | 258.23 g/mol | C10H14N2O6 | 1 G
SDP
Catalog No. 5000301356
Encompass_Preferred
2'-C-methyluridine is a uridine nucleoside analog supplied as a white to off-white solid for laboratory research. It is used in biochemical and pharmacological studies of nucleoside analogs and as a reagent in enzymatic and antiviral assays.
  • Provides a uridine analog for nucleoside research.
  • High purity suitable for analytical and biochemical applications.
  • Soluble in water; may require ultrasonic agitation for complete dissolution.
  • Stable when stored as a powder under low-temperature conditions.
  • Available in small laboratory sizes for research workflows.
Catalog No. 50-003-01356 Supplier Medchemexpress LLC Supplier No. HYW0200981G
2'-C-methyluridine is a uridine nucleoside analog supplied as a white to off-white solid for laboratory research. It is used in biochemical and pharmacological studies of nucleoside analogs and as a reagent in enzymatic and antiviral assays.
  • Provides a uridine analog for nucleoside research.
  • High purity suitable for analytical and biochemical applications.
  • Soluble in water; may require ultrasonic agitation for complete dissolution.
  • Stable when stored as a powder under low-temperature conditions.
  • Available in small laboratory sizes for research workflows.