Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-C-methyluridine | 31448-54-1 | MFCD02682960 | ≥97.0% | 258.23 g/mol | C10H14N2O6 | 5 G
SDP

Catalog No. 5000300467
Encompass_Preferred

2'-C-methyluridine is a modified uridine nucleoside used in biochemical and pharmacological research as a nucleoside analog. It is provided as a stable powder for experimental use in studies of nucleoside metabolism, antiviral activity, and medicinal chemistry.

  • High purity (≥97.0%).
  • Molecular formula C10H14N2O6; molecular weight 258.23 g/mol.
  • Supplied as a white to off-white powder for easy handling.
  • Recommended storage: powder -20°C long-term, 4°C short-term; in solvent -80°C for extended storage.
  • Suitable for biochemical, antiviral, and medicinal chemistry applications.
  • Available in multiple pack sizes to support small-scale and larger studies.

Catalog No. 50-003-00467 Supplier Medchemexpress LLC Supplier No. HYW0200985G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-C-methyluridine is a modified uridine nucleoside used in biochemical and pharmacological research as a nucleoside analog. It is provided as a stable powder for experimental use in studies of nucleoside metabolism, antiviral activity, and medicinal chemistry.

  • High purity (≥97.0%).
  • Molecular formula C10H14N2O6; molecular weight 258.23 g/mol.
  • Supplied as a white to off-white powder for easy handling.
  • Recommended storage: powder -20°C long-term, 4°C short-term; in solvent -80°C for extended storage.
  • Suitable for biochemical, antiviral, and medicinal chemistry applications.
  • Available in multiple pack sizes to support small-scale and larger studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.