Learn More
Medchemexpress LLC 2-carboxybenzaldehyde (phthalaldehydic acid) | 119-67-5 | MFCD00003336 | 50g
SDP
Catalog No. 5000266252
Encompass_Preferred
2-Carboxybenzaldehyde is a key intermediate metabolite in the biodegradation of polycyclic aromatic hydrocarbons (PAHs) such as fluoranthene and phenanthrene 2-Carboxybenzaldehyde can be produced by the degradation of fluoranthene by Pasteurella sp IFA and Mycobacterium sp PYR-1 2-Carboxybenzaldehyde can also be produced by the degradation of phenanthrene by Pseudomonas sp PPD[1][2][3]
Catalog No. 50-002-66252 Supplier Medchemexpress LLC Supplier No. HYY177150G
2-Carboxybenzaldehyde is a key intermediate metabolite in the biodegradation of polycyclic aromatic hydrocarbons (PAHs) such as fluoranthene and phenanthrene 2-Carboxybenzaldehyde can be produced by the degradation of fluoranthene by Pasteurella sp IFA and Mycobacterium sp PYR-1 2-Carboxybenzaldehyde can also be produced by the degradation of phenanthrene by Pseudomonas sp PPD[1][2][3]