Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Carboxyethyl acrylate

Catalog No. 55234850ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-234-850ML 50 mL
55-234-8500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-234-850ML Supplier Sigma Aldrich Supplier No. 55234850ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Can be polymerized in solution or emulsion to produce acrylic, vinyl-acrylic, or styrenic-acrylic polymers with greater flexibility due to lower glass transition of itshomopolymer (<30°C) and improved adhesion.

Specifications

CAS 24615-84-7
Density 1.214 g/mL (at 25°C)
Boiling Point 103°C (19 mmHg)
Molecular Formula C6H8O4
Refractive Index n20/D 1.457 (literature)
Linear Formula CH2=CHCO2(CH2)2 CO2H
Quantity 50 mL
MDL Number MFCD00040709
Synonym beta-(Acryloyloxy)propionic acid; beta-Carboxyethyl acrylate; 3-(Acryloyloxy)propionic acid; 3-Acryloyloxypropanoic acid; Acrylic acid 2-carboxyethyl ester; Acrylic acid dimer
Molecular Weight (g/mol) 144.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.