Learn More
Medchemexpress LLC 2-Carboxyphenol-d4 | 78646-17-0 | 99.8% | 142.15 g·mol⁻¹ | C7H2D4O3 | 1 MG
SDP
Catalog No. 5000300266
Encompass_Preferred
2-Carboxyphenol-d4 is a deuterium-labeled analogue of 2-carboxyphenol intended for use as a tracer or internal standard in quantitative analytical methods. The d4 labeling improves accuracy for isotope-dilution experiments and facilitates detection by NMR, GC-MS, and LC-MS. The compound is provided in milligram-scale packages with high reported purity.
  • Deuterium-labeled (d4) for isotopic tracing
  • High reported purity (99.83%)
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS
  • Molecular formula C7H2D4O3; molecular weight 142.15 g·mol⁻¹
  • Supplied in milligram-scale packages for analytical use
Catalog No. 50-003-00266 Supplier Medchemexpress LLC Supplier No. HYW0200051MG
2-Carboxyphenol-d4 is a deuterium-labeled analogue of 2-carboxyphenol intended for use as a tracer or internal standard in quantitative analytical methods. The d4 labeling improves accuracy for isotope-dilution experiments and facilitates detection by NMR, GC-MS, and LC-MS. The compound is provided in milligram-scale packages with high reported purity.
  • Deuterium-labeled (d4) for isotopic tracing
  • High reported purity (99.83%)
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS
  • Molecular formula C7H2D4O3; molecular weight 142.15 g·mol⁻¹
  • Supplied in milligram-scale packages for analytical use