Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-carboxyphenol-d4 | 78646-17-0 | MFCD06658892 | 99.8% | 142.15 g·mol⁻¹ | C7H2D4O3 | 5 MG
SDP

Catalog No. 5000301811
Encompass_Preferred

2-Carboxyphenol-d4 is the deuterium-labeled analogue of 2-carboxyphenol supplied as an isotopically labeled standard for analytical and biochemical studies. It is suitable for use in mass spectrometry, metabolic tracing, and mechanistic investigations, and is provided with specified purity and handling and storage guidance.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (99.83%).
  • Molecular weight 142.15 g·mol⁻¹; formula C7H2D4O3.
  • Soluble in DMSO at 50 mg/mL; ultrasonic assistance may be required.
  • Store sealed at 4°C; in solvent, store at -80°C up to 6 months.

Catalog No. 50-003-01811 Supplier Medchemexpress LLC Supplier No. HYW0200055MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Carboxyphenol-d4 is the deuterium-labeled analogue of 2-carboxyphenol supplied as an isotopically labeled standard for analytical and biochemical studies. It is suitable for use in mass spectrometry, metabolic tracing, and mechanistic investigations, and is provided with specified purity and handling and storage guidance.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (99.83%).
  • Molecular weight 142.15 g·mol⁻¹; formula C7H2D4O3.
  • Soluble in DMSO at 50 mg/mL; ultrasonic assistance may be required.
  • Store sealed at 4°C; in solvent, store at -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.