Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Carboxyphenyl phosphate, 98%

Catalog No. AAA1441706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1441706 5 g
AAA1441703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1441706 Supplier Thermo Scientific Chemicals Supplier No. A1441706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6064-83-1 | C7H7O6P | 218.101 g/mol

2-Carboxyphenyl phosphate is used as a substrate for the estimation of acid phosphatase. It is also used as a analgesic, anti-inflammatory agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Carboxyphenyl phosphate is used as a substrate for the estimation of acid phosphatase. It is also used as a analgesic, anti-inflammatory agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6064-83-1
Molecular Formula C7H7O6P
Molecular Weight (g/mol) 218.101
MDL Number MFCD00042645
InChI Key FFKUDWZICMJVPA-UHFFFAOYSA-N
Synonym fosfosal, 2-phosphonooxy benzoic acid, disdolen, salicyl phosphate, o-carboxyphenyl phosphate, 2-carboxyphenyl phosphate, 2-phosphonoxybenzoic acid, phosphonoxybenzoic acid, o-phosphonooxy benzoic acid, fosfosal inn
PubChem CID 3418
IUPAC Name 2-phosphonooxybenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)OP(=O)(O)O

Specifications

Melting Point 162°C to 166°C
Quantity 5 g
Beilstein 2619142
Merck Index 14,4255
Solubility Information Soluble in water.
Formula Weight 218.1
Percent Purity 98%
Chemical Name or Material 2-Carboxyphenyl phosphate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 227-993-5
RTECSNumber VO2770000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.