Learn More
Andwin Scientific 2" Cardboard Freezer Boxes, 12/pk
SDP
Catalog No. 50727572
Encompass_Preferred
Freezer Box Only. For use with Stainless Steel Freezer Racks. Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes. Box footprint is 133 x 133 mm. Freezer Box Only. Customers wishing to order freezer box with dividers please use Fisher part numbers 50820929 or NC9804321.
Catalog No. 50-727-572 Supplier Andwin Scientific Supplier No. 6161565
Freezer Box Only. For use with Stainless Steel Freezer Racks. Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes. Box footprint is 133 x 133 mm. Freezer Box Only. Customers wishing to order freezer box with dividers please use Fisher part numbers 50820929 or NC9804321.