Learn More
Alkali Scientific ASI™ 2" Cardboard Freezer Boxes with NO Drain Holes and Low Profile Lid -Water and Ice Resistant Coating, 25 Cell Divider
SDP
Catalog No. 503026841
Encompass_Preferred
Cryo-freezer boxes are protected from liquid nitrogen or condensation from freezers by a moisture repellent coating. Our freezer boxes are made of a sturdy white cardbard and fit all standard-stainless steel and polycarbonate storage systems. For use in mechanical freezers and liquid nitrogen tanks at temperature to -196°C. White water repellent cardboard box for storage of samples or tubes. With Low Profile Lid With Cell Dividers, with NO drain holes Dimensions (L x W x H)   5 5/16 x 5 5/16 x 1 15/16”  
Catalog No. 50-302-6841 Supplier Alkali Scientific Supplier No. FB2CC25LP
Cryo-freezer boxes are protected from liquid nitrogen or condensation from freezers by a moisture repellent coating. Our freezer boxes are made of a sturdy white cardbard and fit all standard-stainless steel and polycarbonate storage systems. For use in mechanical freezers and liquid nitrogen tanks at temperature to -196°C. White water repellent cardboard box for storage of samples or tubes. With Low Profile Lid With Cell Dividers, with NO drain holes Dimensions (L x W x H)   5 5/16 x 5 5/16 x 1 15/16”  