Learn More
Lascar Electronics Inc Dual Channel High Accuracy Thermistor 21CFR WiFi Sensor
SDP
Catalog No. NC1856668
Encompass_Preferred
This 21CFR WiFi-enabled dual channel data logger can remotely monitor an environment using two high accuracy thermistor probes.
Catalog No. NC1856668 Supplier Lascar Electronics Inc Supplier No. ELWIFI21CFRDTP+
This 21CFR WiFi-enabled dual channel data logger can remotely monitor an environment using two high accuracy thermistor probes.