Learn More
Checkers 2-Channel Linebacker™ Cable Protector
Catalog No. 19088470 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Yellow, Orange
Catalog No. Color
19-088-470 Yellow, Orange
1 options
Catalog No. 19-088-470 Supplier Checkers Supplier No. CP2X325Y/O
Only null left
Add to Cart
Add to Cart
Heavy Duty cable protector with hinged lid and wide channel design provides a safer crossing for vehicles and pedestrian traffic while protecting valuable electrical cables and hose lines from damage.
  • Protects cables and hoses up to 3.25 in. outside dia.
  • Patented T-Connectors to extend to any length
  • Reinforced hinged lid for easy cable placement
  • All-weather polyurethane construction
  • Patented 5-bar tread surface for maximum traction
  • Easy to transport, setup, disassemble, and store
  • Ideal for use in oil and gas applications, mining, military, and other industrial environments with heavy trucks and forklift
Specifications
Material Polyurethane
Color Yellow, Orange
Product Type Cable Protector
Dimensions (L x W x H) 39 x 21.75 x 5 in.