Learn More
Checkers 2-Channel Rubber Duct Internet Cable Protector
Catalog No. 19088443 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
19-088-443 Black
1 options
Catalog No. 19-088-443 Supplier Checkers Supplier No. RFD210
Add to Cart
Add to Cart
Rubber duct systems protect and hide valuable cords and cables located in walkways or work areas. These non-metallic, flexible, industrial-grade rubber protectors are extremely durable, easy to install and organize portable cords and cables while reducing trip hazards.
  • Two-Channel Protector Multiple Communication Lines Or 14 Through 22 Gauge Wire
  • Can Be Slit If Necessary (Supplied Unslit)
  • Reduces Tripping Hazards
  • Protects And Organizes Portable Cords And Cables
  • Non-Metallic, Industrial-Grade Rubber Construction
  • Multiple Sizes For Various Cable Diameters
  • Grooved Underside For Better Traction
  • Optional Connector Kit Available For Most Sizes
Specifications
Material Rubber
Color Black
Length (English) 10 ft.
Length (Metric) 3 m
Product Type Cable Protector