Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Two-Channel Protector Multiple Communication Lines Or 14 Through 22 Gauge Wire
- Can Be Slit If Necessary (Supplied Unslit)
- Reduces Tripping Hazards
- Protects And Organizes Portable Cords And Cables
- Non-Metallic, Industrial-Grade Rubber Construction
- Multiple Sizes For Various Cable Diameters
- Grooved Underside For Better Traction
- Optional Connector Kit Available For Most Sizes
Specifications
Specifications
| Material | Rubber |
| Product Type | Cable Protector |
| Length (English) | 10 ft. |
| Length (Metric) | 3 m |
| Color | Black |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?