Learn More
Alkali Scientific ASI™ 2 Channel Sloping Bottom Reagent Solution Reservoir, 25mL, 1/pk, 50/ Case
SDP
Catalog No. 503027507
Encompass_Preferred
25 mL Reagent Soultion Reservoir, unique 2 channel column basins, PS, White, V bottom, Sterile The Solution Reservoir, 25mL, with its innovative 2 channel column basins, exemplifies a commitment to quality and functionality. Whether used in research, analysis, or experimentation, streamline your laboratory processes and elevate your work with this cutting-edge solution reservoir. 
Catalog No. 50-302-7507 Supplier Alkali Scientific Supplier No. RS831
25 mL Reagent Soultion Reservoir, unique 2 channel column basins, PS, White, V bottom, Sterile The Solution Reservoir, 25mL, with its innovative 2 channel column basins, exemplifies a commitment to quality and functionality. Whether used in research, analysis, or experimentation, streamline your laboratory processes and elevate your work with this cutting-edge solution reservoir. 