Learn More
Checkers 2-Channel Yellow Jacket™ Extreme Heavy-Duty Cable Protector
Catalog No. 19088403 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Black on Yellow
Catalog No. Color
19-088-403 Black on Yellow
1 options
Catalog No. 19-088-403 Supplier Checkers Supplier No. YJ2400Y/B
Only null left
Add to Cart
Add to Cart
Durable urethane construction withstands the load of a 400-ton haul truck and endures the constant pounding, impact and abrasion caused by haul trucks and falling rocks. Modular interlocking design is ideal for use in industrial and heavy duty applications.
  • Protects cables and hoses up to 3.75 in. outside dia.
  • Dog bone connectors to extend to any desired length
  • Thick lid and base allow for high volume of traffic
  • Hinged lid for easy cable placement
  • Modular interlocking design
  • Tested to with stand a fully loaded 400-ton haul truck
Specifications
Color Black on Yellow
Material Polyurethane
Product Type Cable Protector