Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-1,1,1-trimethoxyethane

Catalog No. 4379485ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-794-85ML 5 mL
43-794-825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-794-85ML Supplier Sigma Aldrich Supplier No. 4379485ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-chloro-1,1,1-trimethoxyethane is a chloromethyl heterocyclic compound
  • It participates in the synthesis of 2-phenyl-1,3,4-oxadiazole derivatives.2-chloro-1,1,1-trimethoxyethane may be used for the synthesis of chlorotriazolinone, an nk1 antagonist and 2-chloromethyl-7-[3-(trifluoromethyl)pyridin-2-yl]-3h-quinazolin-4-one

Specifications

CAS 74974-54-2
Density 1.147 g/mL (at 25°C (literature))
Boiling Point 138°C (lit.)
Molecular Formula C5H11ClO3
Refractive Index n20/D 1.425 (literature)
Linear Formula ClCH2C(OCH3)3
MDL Number MFCD00216629
Quantity 5 mL
Synonym Trimethyl chloro-orthoacetate
Molecular Weight (g/mol) 154.59
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.