Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-1-[2-[3-(trifluoromethyl)anilino]phenyl]ethanone | 2378626-29-8 | 99.0% | 313.70 g/mol | C15H11ClF3NO | 10MG
SDP

Catalog No. 5000236628
Encompass_Preferred

TED-347 is a potent, irreversible, covalent, allosteric small-molecule inhibitor of the YAP-TEAD protein-protein interaction that blocks TEAD transcriptional activity and has demonstrated antitumor activity in reported assays.

  • Irreversible covalent inhibitor of YAP-TEAD interaction (EC50 = 5.9 μM for TEAD4*Yap1).
  • Covalently binds Cys-367 in TEAD4 (Ki = 10.3 μM).
  • Molecular formula C15H11ClF3NO; molecular weight 313.70 g/mol.
  • CAS number 2378626-29-8.
  • High purity (98.97%).
  • Appearance: light yellow to green yellow solid.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (2 years) or -20 °C (1 year).

Catalog No. 50-002-36628 Supplier Medchemexpress LLC Supplier No. HY12526910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TED-347 is a potent, irreversible, covalent, allosteric small-molecule inhibitor of the YAP-TEAD protein-protein interaction that blocks TEAD transcriptional activity and has demonstrated antitumor activity in reported assays.

  • Irreversible covalent inhibitor of YAP-TEAD interaction (EC50 = 5.9 μM for TEAD4*Yap1).
  • Covalently binds Cys-367 in TEAD4 (Ki = 10.3 μM).
  • Molecular formula C15H11ClF3NO; molecular weight 313.70 g/mol.
  • CAS number 2378626-29-8.
  • High purity (98.97%).
  • Appearance: light yellow to green yellow solid.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (2 years) or -20 °C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.