Learn More
Medchemexpress LLC 2-chloro-1-[2-[3-(trifluoromethyl)anilino]phenyl]ethanone | 2378626-29-8 | 99.0% | 313.70 g/mol | C15H11ClF3NO | 25MG
SDP
Catalog No. 5000238240
Encompass_Preferred
TED-347 is a small-molecule, irreversible, covalent and allosteric inhibitor of the YAP-TEAD protein-protein interaction. It covalently modifies TEAD4 at Cys-367, blocks TEAD transcriptional activity, and shows cellular antitumor activity (EC50 5.9 μM; Ki 10.3 μM). The compound is supplied as a solid with high purity for research use only.
  • Irreversible covalent inhibitor of YAP-TEAD interaction.
  • Targets TEAD4 Cys-367 via covalent modification.
  • Demonstrated cellular activity (EC50 5.9 μM; Ki 10.3 μM).
  • High purity suitable for research applications.
  • Available in multiple small pack sizes for experimental workflows.
Catalog No. 50-002-38240 Supplier Medchemexpress LLC Supplier No. HY12526925MG
TED-347 is a small-molecule, irreversible, covalent and allosteric inhibitor of the YAP-TEAD protein-protein interaction. It covalently modifies TEAD4 at Cys-367, blocks TEAD transcriptional activity, and shows cellular antitumor activity (EC50 5.9 μM; Ki 10.3 μM). The compound is supplied as a solid with high purity for research use only.
  • Irreversible covalent inhibitor of YAP-TEAD interaction.
  • Targets TEAD4 Cys-367 via covalent modification.
  • Demonstrated cellular activity (EC50 5.9 μM; Ki 10.3 μM).
  • High purity suitable for research applications.
  • Available in multiple small pack sizes for experimental workflows.