Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-1,3,2-dioxaphospholane 2-oxide

Catalog No. 3779535G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-795-35G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-795-35G Supplier Sigma Aldrich Supplier No. 3779535G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chloro-1,3,2-dioxaphospholane 2-oxide

Specifications

CAS 6609-64-9
Melting Point 12°C to 14°C (neat) (lit.)
Density 1.55 g/mL (at 25°C (literature))
Boiling Point 89°C to 91°C (0.8 mmHg)
Molecular Formula C2H4ClO3P
Refractive Index n20/D 1.45 (literature)
Linear Formula C2H4ClO3P
MDL Number MFCD00043138
Quantity 100 mg
Synonym 2-Chloro-2-oxo-1,3,2-dioxaphospholane; Ethylene glycol chlorophosphate
Molecular Weight (g/mol) 142.48
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.