Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-1,3,2-Dioxaphospholane

Catalog No. 3912205ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-122-05ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 39-122-05ML Supplier Sigma Aldrich Supplier No. 3912205ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphitylating agent used in the synthesis of phosphocholines and spirophosphoranes.

Specifications

CAS 75507-26-5
Density 1.192 g/mL at 25°C
Boiling Point 115°C (0.04 mmHg, lit.)
Molecular Formula C9H18O5
Refractive Index n20/D 1.489
Linear Formula C9H18O5
MDL Number MFCD00067524
Quantity 5 mL
Synonym Cyclic ethylene chlorophosphite; Cyclic ethylene phosphorochloridite; Ethylene chlorophosphite; Ethylene glycol chlorophosphite
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.