Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-1,3-Bis(2,6-Diisopropylphenyl)Imidazolium Chloride

Catalog No. 904848500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-484-8500MG 100 g
90-484-82G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-484-8500MG Supplier Sigma Aldrich Supplier No. 904848500MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This chloroimidazolium chloride is used in the deoxyfluorination of phenols. Recently, the Ritter lab used it in conjunction with 902314 and 906794 to incorporate the [18F]fluoride radiolabel site specifically into polypeptides for use with positron emission tomography (PET) imaging in the study of various biological applications.Site-specific deoxyfluorination of small peptides with [18F]fluorideHDAC6 Brain Mapping with [18F]Bavarostat Enabled by a Ru-Mediated Deoxyfluorination18F-Deoxyfluorination of Phenols via Ru p-Complexes

Specifications

CAS 368-39-8
Molecular Formula C6H15BF4O
Linear Formula (C2H5)3O(BF4)
MDL Number MFCD00044423
Quantity 100 g
Molecular Weight (g/mol) 189.99 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.