Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,3-difluoro-4-nitrobenzene 98.0+%, TCI America™
SDP

Catalog No. C29645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29645G Supplier TCI America Supplier No. C29645G
Add to Cart
Add to Cart

Specifications

CAS 3847-58-3
Melting Point 47°C
Color Yellow
Molecular Formula C6H2ClF2NO2
Quantity 5 g
MDL Number MFCD00042201
UN Number 1759
Synonym 3-chloro-2,4-difluoronitrobenzene, 2,4-difluoro-3-chloronitrobenzene, benzene, 2-chloro-1,3-difluoro-4-nitro, 2,4-difluoro-3-chloronitrobnzene, 3-chloro-2,4-difluoro-1-nitrobenzene, 2-chloro-1,3-difluoro-4-nitro-benzene, pubchem4320, acmc-209j0b, ksc911k8t, buttpark 177\01-33
InChI Key CTVBVNKEMCGZDF-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1[N+](=O)[O-])F)Cl)F
IUPAC Name 2-chloro-1,3-difluoro-4-nitrobenzene
Molecular Weight (g/mol) 193.534
PubChem CID 223080
Formula Weight 193.53
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1,3-difluoro-4-nitrobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.