Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,3-difluorobenzene 98.0+%, TCI America™
SDP

Catalog No. C22115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22115G Supplier TCI America Supplier No. C22115G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38361-37-4
Molecular Formula C6H3ClF2
Molecular Weight (g/mol) 148.537
MDL Number MFCD00142604
InChI Key OTZQYBFTOANOJO-UHFFFAOYSA-N
PubChem CID 142257
IUPAC Name 2-chloro-1,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)F)Cl)F

Specifications

Color Yellow
Boiling Point 131°C
Quantity 5 g
UN Number 1993
Formula Weight 148.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-1,3-difluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.