Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,3-dimethylimidazolinium Chloride (ca. 25% in Dichloromethane), TCI America™
SDP

Catalog No. C163925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C163925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C163925G Supplier TCI America Supplier No. C163925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37091-73-9
Molecular Formula C5H10Cl2N2
Molecular Weight (g/mol) 169.05
MDL Number MFCD09039290
InChI Key AEBBXVHGVADBHA-UHFFFAOYSA-M
Synonym 2-chloro-1,3-dimethylimidazolinium chloride, unii-56ka2a1u1d, 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium chloride, dmc mi, 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium chloride, pubchem16443, 2-chloro-1,3-dimethyl-4,5-dihydro-3h-imidazol-1-ium chloride, 1h-imidazolium, 2-chloro-4,5-dihydro-1,3-dimethyl-, chloride 1:1, 2-chloro-1,3-dimethylimidazolidin-2-ylium chloride
PubChem CID 10176306
IUPAC Name 2-chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium chloride
SMILES [Cl-].CN1CC[N+](C)=C1Cl

Specifications

Quantity 25 g
UN Number 1593
Formula Weight 169.05
Physical Form Liquid
Chemical Name or Material 2-Chloro-1,3-dimethylimidazolinium Chloride (ca. 25% in Dichloromethane)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.