Learn More
AdipoGen 2-Chloro-1 3-propanediol
SDP
Supplier:  AdipoGen CDXC0752M025
Encompass_Preferred
Chemical. CAS 497-04-1. Formula C3H7ClO2. MW BD9837. 2-Chloro-1,3-propanediol 2-MCPD is a heat-induced food contaminant present in oil- and fat-containing foodstuff. 2-MCPD is formed during the refinement of vegetable oils and fats and is therefore present in numerous oil- and fat-containing foods. This compound can be used as a reference compound for analytical studies.
Catalog No. 50-256-1344