Learn More
Selleck Chemical LLC 2-Chloro-1-(4-fluorobenzyl)benzimidazole S5446-25mg
SDP
Catalog No. 503109217
Encompass_Preferred
2-Chloro-1-(4-fluorobenzyl)benzimidazole is an aldose reductase (ALR2) inhibitor which is used for preparation of disubstituted benzimidazole derivative
Catalog No. 50-310-9217 Supplier Selleck Chemical LLC Supplier No. S544625MG
2-Chloro-1-(4-fluorobenzyl)benzimidazole is an aldose reductase (ALR2) inhibitor which is used for preparation of disubstituted benzimidazole derivative