Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-1-(4-methoxy-1H-indol-3-yl)ethanone | 858752-72-4 | 98.2% | 223.66 g/mol | C11H10ClNO2 | 1 G
SDP

Catalog No. 5000210106
Encompass_Preferred

2-Chloro-1-(4-methoxy-1H-indol-3-yl)ethanone is an indole-derived chloroethanone used as a drug intermediate in the synthesis of biologically active compounds.

  • cas number: 858752-72-4.
  • molecular formula: C11H10ClNO2.
  • molecular weight: 223.66 g/mol.
  • appearance: solid.
  • purity (lcms): 98.17% (coa).
  • recommended storage: powder: -20°C (3 years), 4°C (2 years); in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-10106 Supplier Medchemexpress LLC Supplier No. HYW0479991G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloro-1-(4-methoxy-1H-indol-3-yl)ethanone is an indole-derived chloroethanone used as a drug intermediate in the synthesis of biologically active compounds.

  • cas number: 858752-72-4.
  • molecular formula: C11H10ClNO2.
  • molecular weight: 223.66 g/mol.
  • appearance: solid.
  • purity (lcms): 98.17% (coa).
  • recommended storage: powder: -20°C (3 years), 4°C (2 years); in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.