Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-1-(4-methoxy-1H-indol-3-yl)ethanone | 858752-72-4 | 98.2% | 223.66 g/mol | C11H10ClNO2 | 100 MG
SDP

Catalog No. 5000210354
Encompass_Preferred

2-chloro-1-(4-methoxy-1H-indol-3-yl)ethanone is an indole-derived chemical intermediate used in the synthesis of biologically active compounds and research reagents. It is supplied as a solid powder for laboratory research and is accompanied by certificate of analysis (COA) and safety data sheet (SDS) documentation.

  • Used as a synthetic intermediate for drug discovery and research.
  • High purity: 98.2% (LCMS).
  • Molecular formula C11H10ClNO2; molecular weight 223.66 g/mol.
  • Physical form: solid powder; recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Safety considerations: harmful if swallowed; causes skin and eye irritation; may cause respiratory irritation.

Catalog No. 50-002-10354 Supplier Medchemexpress LLC Supplier No. HYW047999100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-chloro-1-(4-methoxy-1H-indol-3-yl)ethanone is an indole-derived chemical intermediate used in the synthesis of biologically active compounds and research reagents. It is supplied as a solid powder for laboratory research and is accompanied by certificate of analysis (COA) and safety data sheet (SDS) documentation.

  • Used as a synthetic intermediate for drug discovery and research.
  • High purity: 98.2% (LCMS).
  • Molecular formula C11H10ClNO2; molecular weight 223.66 g/mol.
  • Physical form: solid powder; recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Safety considerations: harmful if swallowed; causes skin and eye irritation; may cause respiratory irritation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.