Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-1-(4-methoxy-1H-indol-3-yl)ethanone | 858752-72-4 | 98.6% | 223.66 g/mol | C11H10ClNO2 | 250 MG
SDP

Catalog No. 5000209601
Encompass_Preferred

2-Chloro-1-(4-methoxy-1H-indol-3-yl)ethanone is an indole-derived chloroacetyl reagent used as a drug intermediate for medicinal chemistry and synthetic organic synthesis. It is supplied as a powder suitable for laboratory research and synthesis.

  • High purity: 98.62%.
  • CAS number: 858752-72-4.
  • Molecular formula: C11H10ClNO2.
  • Molecular weight: 223.66 g/mol.
  • Available pack sizes include 250 mg among others.
  • Storage: powder - -20°C (3 years) or 4°C (2 years); in solvent - -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-09601 Supplier Medchemexpress LLC Supplier No. 250MGHYW047999
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloro-1-(4-methoxy-1H-indol-3-yl)ethanone is an indole-derived chloroacetyl reagent used as a drug intermediate for medicinal chemistry and synthetic organic synthesis. It is supplied as a powder suitable for laboratory research and synthesis.

  • High purity: 98.62%.
  • CAS number: 858752-72-4.
  • Molecular formula: C11H10ClNO2.
  • Molecular weight: 223.66 g/mol.
  • Available pack sizes include 250 mg among others.
  • Storage: powder - -20°C (3 years) or 4°C (2 years); in solvent - -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.