Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone | 30030-91-2 | MFCD03289190 | 223.66 | C11H10ClNO2 | 500 MG
SDP

Catalog No. 5000210107
Encompass_Preferred

2-Chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone is an indole-derived chloro ethanone supplied as a research chemical intermediate for synthesis of active compounds. It is provided in multiple package sizes for laboratory use and is intended for research and drug-development purposes only.

  • Used as a drug intermediate for synthesis of various active compounds.
  • CAS number: 30030-91-2.
  • Molecular formula: C11H10ClNO2, molecular weight: 223.66.
  • Available in multiple package sizes including 500 MG.
  • Intended for research and drug-development use; not for individual or clinical use.

Catalog No. 50-002-10107 Supplier Medchemexpress LLC Supplier No. HYW048171500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone is an indole-derived chloro ethanone supplied as a research chemical intermediate for synthesis of active compounds. It is provided in multiple package sizes for laboratory use and is intended for research and drug-development purposes only.

  • Used as a drug intermediate for synthesis of various active compounds.
  • CAS number: 30030-91-2.
  • Molecular formula: C11H10ClNO2, molecular weight: 223.66.
  • Available in multiple package sizes including 500 MG.
  • Intended for research and drug-development use; not for individual or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.