Learn More
Medchemexpress LLC 2-chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone | 30030-91-2 | MFCD03289190 | 223.66 g·mol⁻¹ | C11H10ClNO2 | 1 G
SDP
Catalog No. 5000210104
Encompass_Preferred
2-Chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone is a research chemical intermediate used in the synthesis of bioactive indole derivatives and other active compounds. It is supplied as a powder for laboratory synthesis and analytical applications.
  • drug intermediate for synthesis of active indole derivatives.
  • molecular formula C11H10ClNO2 and molecular weight 223.66 g·mol⁻¹.
  • available in multiple sizes including 1 g for small-scale research.
  • powder form, stored and shipped at ambient conditions unless otherwise specified.
  • analytical data available (HNMR, CNMR, HPLC, LC-MS) to support purity assessment.
  • hazard: irritant; handle with appropriate personal protective equipment and follow safety data sheet guidance.
Catalog No. 50-002-10104 Supplier Medchemexpress LLC Supplier No. HYW0481711G
2-Chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone is a research chemical intermediate used in the synthesis of bioactive indole derivatives and other active compounds. It is supplied as a powder for laboratory synthesis and analytical applications.
  • drug intermediate for synthesis of active indole derivatives.
  • molecular formula C11H10ClNO2 and molecular weight 223.66 g·mol⁻¹.
  • available in multiple sizes including 1 g for small-scale research.
  • powder form, stored and shipped at ambient conditions unless otherwise specified.
  • analytical data available (HNMR, CNMR, HPLC, LC-MS) to support purity assessment.
  • hazard: irritant; handle with appropriate personal protective equipment and follow safety data sheet guidance.