Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone | 30030-91-2 | MFCD03289190 | 95.0% | 223.66 g/mol | C11H10ClNO2 | 5 G
SDP

Catalog No. 5000210357
Encompass_Preferred

2-chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone is a chemical intermediate used in the synthesis of pharmaceutical and bioactive compounds.

  • Used as a drug intermediate in medicinal chemistry syntheses.
  • Contains a 5-methoxyindole core and a chloro-ethanone functionality.
  • Molecular formula C11H10ClNO2; molecular weight 223.66 g/mol.
  • Supplied in small laboratory quantities for research use.
  • Handle with appropriate laboratory safety precautions.

Catalog No. 50-002-10357 Supplier Medchemexpress LLC Supplier No. HYW0481715G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone is a chemical intermediate used in the synthesis of pharmaceutical and bioactive compounds.

  • Used as a drug intermediate in medicinal chemistry syntheses.
  • Contains a 5-methoxyindole core and a chloro-ethanone functionality.
  • Molecular formula C11H10ClNO2; molecular weight 223.66 g/mol.
  • Supplied in small laboratory quantities for research use.
  • Handle with appropriate laboratory safety precautions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.