Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone | 30030-91-2 | MFCD03289190 | 97.0% | 223.66 g·mol-1 | C11H10ClNO2 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHYW048171

Encompass_Preferred

2-Chloro-1-(5-methoxy-1H-indol-3-yl)-ethanone is an indole-derived organic intermediate used in medicinal chemistry for the synthesis of various active compounds. It is supplied in laboratory-scale quantities for research use and is characterized by molecular formula C11H10ClNO2 and molecular weight 223.66 g·mol-1.

  • Used as a synthetic intermediate in drug development
  • CAS number 30030-91-2
  • Molecular formula C11H10ClNO2
  • Molecular weight 223.66 g·mol-1
  • Available in laboratory-scale package sizes (100 mg-50 g)
  • Intended for research use only

Catalog No. 50-002-09682


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.