Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-1-(6-Methoxy-1 2 3 4-Tetrahydroquinolin-1-Yl)Ethan-1-One

Catalog No. 912131100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-213-1100MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-213-1100MG Supplier Sigma Aldrich Supplier No. 912131100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chloro-1-(6-methoxy-1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one is a cysteine-reactive small-molecule fragment for chemoproteomic and ligandability studies for both traditionally druggable proteins as well as undruggable, or difficult-to-target, proteins. This fragment electrophile, or scout fragment, can be used alone in fragment-based covalent ligand discovery or incorporated into bifunctional tools such as electrophilic PROTAC™ molecules for targeted protein degradation as demonstrated by the Cravatt Lab Lab for E3 ligase discovery.Technology Spotlight: Proteomic Ligandability AssessmentThe Proteome-Wide Potential for Reversible Covalency at CysteineElectrophilic PROTACs that degrade nuclear proteins by engaging DCAF16Proteome-wide covalent ligand discovery in native biological systems PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 194659-03-5
Melting Point >300°C
Molecular Formula C80H80N4O16Rh2
Linear Formula C80H80N4O16Rh2
Quantity 250 mg
Synonym Tetrakis[(S)-(+)-(1-adamantyl)-(N-phthalimido)acetato]dirhodium(II)
Molecular Weight (g/mol) 1559.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.