Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-2-Chloro-1-Phenylethanol

Catalog No. 3635611ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-356-11ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-356-11ML Supplier Sigma Aldrich Supplier No. 3635611ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(-)-2-Chloro-1-Phenylethanol

Specifications

CAS 955-40-8
Density 1.048 g/mL at 25°C
Molecular Formula C14H19NO2
Linear Formula C14H19NO2
Refractive Index n20/D 1.552
Quantity 5 g
MDL Number MFCD00075531
Synonym (R)-(-)-alpha-(Chloromethyl)benzenemethanol
Molecular Weight (g/mol) 233.31 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.